ContactUsLinkComponent
StemFate™ CFC/CFU Differentiation Assay Kit for Human Cells VI, BioVision, Inc.
StemFate™ CFC/CFU Differentiation Assay Kit for Human Cells VI, BioVision, Inc.
  76200-082
 :  Avantor
 :  
undefined
StemFate™ CFC/CFU Differentiation Assay Kit for Human Cells VI, BioVision, Inc.
  76200-082
 :  Avantor
 :  
  • Description:
    StemFate™-4 CFC/CFU Differentiation Assay Kit for Human Cells VI
  • Size:
    50 assays
  • Cat. No.:
    76200-082
Simple, complete stem cell differentiation assay kit.
  • CFUs are formed in semi-solid methylcellulose
Target populations: Granulocyte and macrophage.
It is a miniaturized, in vitro colony-forming cell (CFC) or unit (CFU) differentiation assay that detects the differentiation ability and/or potential of lympho-hematopoietic stem, progenitor or precursor cells to form colonies in semi-solid methylcellulose.
StemfateTM-4 assay utilizes specially designed 35mm petri dishes containing 4 mini-wells (0.1 ml each) which allow colonies to be easily viewed, differentiated and counted up to the edge of the well. The StemFateTM-4 assay kit includes a superior Master Mix to support optimal growth and enumeration of your lympho-hematopoietic cell population of interest, including erythroid progenitors (CFU-E and BFU-E), granulocyte/macrophage progenitors (CFC-GM, CFU-M, CFU-G), multi-potential granulocyte, erythroid, macrophage and megakaryocyte progenitors (CFU-GEMM), and more in just 10 to 14 days.
 : Includes 35 mm petri dishes (4 wells)