Order Entry
United States
ContactUsLinkComponent
PeproGrow™ HEK293 Media, Peprotech, Inc.
PeproGrow™ HEK293 Media, Peprotech, Inc.
Catalog # 10778-850
Supplier:  Avantor
CAS Number:  
PeproGrow™ HEK293 Media, Peprotech, Inc.
Catalog # 10778-850
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1 L
  • Environmentally Preferable:
  • Storage Temperature:
    See product insert for storage instructions.
  • Cat. No.:
    10778-850
  • Supplier no.:
    AFCDHEK293

Specifications

About this item

PeproTech's PeproGrow™ HEK293 is an animal component-free, protein-free, serum-free, chemically-defined, complete medium formulation for the in vitro cultivation of HEK293 cells.

  • Complete Media
  • Animal component-Free
  • Serum-Free
  • Protein-Free
  • Chemically-Defined

PeproGrow™ HEK293 is an animal component-free, protein-free, serum-free, chemically-defined, complete medium formulation for the in vitro cultivation of HEK293 cells (Thermo Fisher Scientific FreeStyle™ 293-F cells, catalog number R790-07). This medium is intended for recombinant protein expression in suspension culture, which is recommended for a 5 day batch culture with a seeding density of 0.6 x106 cells/mL. This ready-to-use medium contains L-alanyl-L-Glutamine, amino acids, vitamins, and salts. An adaptation process is not required for Thermo Fisher Scientific FreeStyle™ 293-F cells.