About this item
InVivoMAb anti-mouse CD200 (OX2), Clone: OX-90, Isotype: rat IgG2a, κ, Applications: in vivo CD200 blockade, in vitro CD200 blockade, Immunohistochemistry (frozen), Immunofluorescence, Flow cytometry
Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Rat
Host:
Isotype:
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76555-172
- 76556-166
- 76556-032
- 76555-812
- 76554-920
- Antigen Symbol:
- CD200
- CD200
- CD200
- CD200
- CD200
- Antigen Synonyms:
- OX2
- OX2
- OX2
- OX2
- OX2
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Host:
- Rat
- Rat
- Rat
- Rat
- Rat