Advanced Chemistry Go Direct™ Package
Catalog # 470303-858
Supplier: Vernier Software & Technology
Specifications
- Interface:
- Description:Advanced chemistry go Direct™ package
- Environmentally Preferable:
- Cat. No.:470303-858
Specifications
About this item
We offer different packages to meet your needs and budget for the Advanced Chemistry classroom.
- Probes include temperature, pH, gas pressure, voltage; conductivity, drop counter, constant current, ORP, and spectrophotometer
The starter package is great way to get started, and the deluxe package includes all the sensors needed for the experiments in the Advanced Chemistry with Vernier Book item 470017-580.