Orders LinkContactUsLinkComponent
Vernier® Go Direct™ Rotary Motion Sensor
Vernier® Go Direct™ Rotary Motion Sensor
Vernier® Go Direct™ Rotary Motion Sensor
Certificates
Vernier® Go Direct™ Rotary Motion Sensor
2 Options Available Below
Product Details & Documents
The Go Direct Rotary Motion Sensor measures angular motion.
  • Determine position, velocity, and acceleration of objects in angular motion
  • Measure angle increments as small as 0.25°
  • Investigate rotational dynamics, conservation of angular momentum, pendulum motion, polarization of light, and more
Monitor angular motion easily and precisely with the Go Direct Rotary Motion Sensor, which connects wirelessly via Bluetooth® or wired via USB to your device. The wireless connection eliminates the cables that can get caught and tangled during rotational investigations.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting-no interruptions or inconvenience to you or your students.
Our Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth wireless technology, allowing you to choose the solution best for your classroom or laboratory.
Delivery information: Includes Go Direct Rotary Motion Sensor, thumb screw, three-step pulley and mounting screw, O-ring.
Specifications for Product Family
Specifications
  • Catalog No:
  • 470359-996
  • 470303-844
  • Interface:
  • USB
  • USB
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 470359-996
    Go Direct™ Rotary motion sensor, with accessory kit
    Go Direct™ Rotary motion sensor, with accessory kit
    Log in for availability
    Log in for availability
    Each
    Each - $340.99
    $340.99
    Each
       $340.99
    Minimum order quantity is 1
    Supplier Number: GDX-RMPK
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    470359-996
    Interface
    USB
    New Product
  • 470303-844
    Go Direct™ Rotary motion sensor
    Go Direct™ Rotary motion sensor
    Log in for availability
    Log in for availability
    Each
    Each - $204.79
    $204.79
    Each
       $204.79
    Minimum order quantity is 1
    Supplier Number: GDX-RMS
    Specifications:
    More
    Specifications
    Cat. No.
    470303-844
    Interface
    USB