ContactUsLinkComponent
Go Direct™ EKG
Go Direct™ EKG
Catalog # 470303-912
undefined
Go Direct™ EKG
Catalog # 470303-912
Supplier Number:  GDX-EKG
Specifications
  • Description:
    Go Direct™ EKG
  • Cat. No.:
    470303-912
Specifications
About this item
The Go Direct EKG Sensor records electrical signals produced during heart or muscle contractions.
  • Compare and measure students’ electrocardiogram (EKG/ECG) waveforms
  • Determine heart rate by examining the number of QRS waveforms in a series of electrocardiograms (EKG/ECGs)
  • Study contractions of muscles (EMG) in the arm, leg, or jaw
  • Correlate measurements of grip strength and electrical activity with muscle fatigue
The Go Direct EKG Sensor measures electrical activity in the heart and electrical signals produced during muscle contractions. The wireless options minimize the concern of cables getting caught and tangled during experiments. This sensor provides two separate outputs: one optimized for standard 3-lead EKG tracings and one optimized for surface EMG recordings. EMG recordings can also be rectified automatically by the sensor.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting-no interruptions or inconvenience to you or your students.
Our Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth wireless technology, allowing you to choose the solution best for your classroom or laboratory.