ContactUsLinkComponent
Go Direct™ CO₂ Gas Sensor
Go Direct™ CO₂ Gas Sensor
Catalog # 470303-908
undefined
Go Direct™ CO₂ Gas Sensor
Catalog # 470303-908
Supplier Number:  GDX-CO2
Specifications
  • Description:
    Go Direct™ CO₂ gas sensor
  • Cat. No.:
    470303-908
Specifications
About this item
The Go Direct CO₂ Gas Sensor measures gaseous carbon dioxide concentration levels, air temperature, and relative humidity.
  • Measure the change in gas concentration during cellular respiration
  • Measure the change in gas concentration during photosynthesis
  • Study the effect of temperature on cell respiration or metabolism of organisms
  • Investigate artificial selection in yeast by comparing respiration rates of different strains
Monitor changes in carbon dioxide, temperature, and relative humidity easily with the Go Direct CO2 Gas Sensor. This sensor includes built-in temperature compensation and humidity protection. A 250 ml Nalgene bottle is included for running controlled experiments with small plants and animals.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting-no interruptions or inconvenience to you or your students.
Our Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth wireless technology, allowing you to choose the solution best for your classroom or laboratory.