Specifications
- Description:MINI-SPIKE® IV Additive Dispensing Pin
- Cat. No.:76202-656
Specifications
About this item
MINI-SPIKE® IV Additive dispensing pin for preparing and dispensing diluent or additive from multi-dose rubber-stoppered vials
- Sterile
- DEHP-free and latex-free