Thermal Papers, ABS Imaging Systems
MSPP-K65HM-CE
- Color:Black/White
- Description:Thermal paper
- Cat. No.:MSPP-K65HM-CE
- Supplier no.:K65HM-CE
High density and high contrast thermal papers for printers.
: Sold by the pack.
: Packed 4 rolls per box, 215 prints per roll.