ContactUsLinkComponent
Phosphoenolpyruvate Carboxykinase Activity Assay Kit (Colorimetric), BioVision Inc.
Phosphoenolpyruvate Carboxykinase Activity Assay Kit (Colorimetric), BioVision Inc.
  76265-732
 :  Avantor
 :  
undefined
Phosphoenolpyruvate Carboxykinase Activity Assay Kit (Colorimetric), BioVision Inc.
  76265-732
 :  Avantor
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Phosphoenolpyruvate Carboxykinase Activity Assay Kit
  • Environmentally Preferable:
  • Cat. No.:
    76265-732
  • No. of assays/kit:
    100 assays
The simple and convenient assay to measure PEPCK activity in tissues and cell lines.
  • High-throughput adaptable, convenient and sensitive
  • Detect as low as 10 µU of PEPCK activity in a variety of samples
Phosphoenolpyruvate Carboxykinase (PEPCK EC 4.1.1.32) is an enzyme which belongs to the lyase family. In the presence of GTP, it catalyzes the reversible conversion of oxaloacetate (OAA) into phosphoenolpyruvate (PEP), GDP and CO2.
In humans, two isoforms of PEPCK are found such as cytosolic form (PEPCK-C, also called PCK1) and mitochondrial form (PEPCK-M). PEPCK-C is a rate-controlling step in gluconeogenesis. Recent studies found abnormal concentrations of PEPCK in diabetic mice. Therefore, accurate measurement of Phosphoenolpyruvate Carboxykinase activity is valuable for both, mechanistic and therapeutic studies.
This Phosphoenolpyruvate Carboxykinase (PEPCK) activity assay kit provides a quick and easy way for measuring PEPCK activity in various samples. In this assay, Phosphoenolpyruvate Carboxylase is coupled with a set of enzymes that convert PEP and carbonate into a series of intermediates and hydrogen peroxide, which in turn, reacts with a probe and converted generating a colorimetric signal (OD: 570 nm). The color intensity is directly proportional to the amount of active Phosphoenolpyruvate Carboxylase present in samples.