Orders LinkContactUsLinkComponent
Bitrex® Sensitivity Solution, Draeger
Bitrex® Sensitivity Solution, Draeger
Sensitivity solution, 4 oz.
Catalog # 34330-906
Supplier:   Draeger
Supplier #:  4055623
Bitrex® Sensitivity Solution, Draeger
Catalog # 34330-906
Supplier:  Draeger
Bitrex® Sensitivity Solution, Draeger
Sensitivity solution, 4 oz.
Price
$35.04
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Fit testing seinsitivity solution for respirators.
  • Replacement solution for the Fit Test Kit
  • Product sprayed in the air to test the fit of face masks
If sprayed, and subject does not taste anything bitter, then mask fits correctly. Presence of a bitter taste, indicates a need for a better fitting mask.
Documents
Specifications
  • Description:
    Sensitivity solution
  • Weight:
    4 oz.