TOC Vials, Sievers Certified
Product Details & Documents
Sievers Certified (<10 ppb) TOC Vials are designed for use in critical pharmaceutical applications such as USP and EP water testing as well as municipal water testing. Certification to below 10 ppb gives you confidence in the quality of TOC measurements.
- Reliable, with guaranteed performance
Scrupulously pre-cleaned in an ISO 9001 environment using a validated, automated process.
Sievers Certified TOC Vials are compatible with most TOC analyzers including Sievers, Shimadzu, OI Analytical, Teledyne-Tekmar and Skalar.
These vials are the right choice for strict regulatory compliance testing.
Documents
Specifications
- Cap included:Yes
- Septum:Teflon-lined
- Capacity:40.00 ml
- Description:TOC Glass Vials with Caps and Septa