Plasticware Starter Set, United Scientific Supplies
Catalog # 76263-320
Supplier: United Scientific Supplies
Specifications
- Description:Plasticware Starter Kit
- Environmentally Preferable:
- Cat. No.:76263-320
Specifications
About this item
A customized plasticware assortment that is designed for general laboratory use.
- Wide range of sizes and varieties
- Translucent design
This plastic starter set includes all the most commonly used items to conduct your experiments without warring about glass breaking.
Delivery: Starter kit includes: 5 Beakers, (50, 100, 250, 500, and 1000 ml, 4 Graduated Cylinders (10, 25, 50, and 100 ml), 2 Wash Bottles (250 and 500 ml), 2 Narrow Mouth Polypropylene Reagent Bottles (250 and 500 ml), 2 Wide Mouth Polypropylene Reagent Bottles (250 and 500 ml), and 2 Measuring Jugs (500 and 1000 ml).