Carrier Trays, United Scientific Supplies
Catalog # 76263-316
Supplier: United Scientific Supplies
Specifications
- Description:Carrier Tray
- Dimensions:15×9.5×4.5"
- Cat. No.:76263-316
Specifications
About this item
General use tray with a built-in handle.
- Easy to grab handle
- Divided section to keep items separate
Steam autoclavable, polypropylene molded tray has a built-in handle for easy carrying of reagents or laboratory ware using one hand.