Ion Transfer Tubes, Thermo Scientific
Supplier: 0000013443
Certificates
About this item
Frequently Bought Together
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
Falcon® Pipette Controller, Corning
Supplier: Corning
AdvanceBio SEC 2.7 μm, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Bead Ruptor Pre-Filled Bead Tubes, 7 ml, Omni International
Supplier: OMNI INTERNATIONAL
ION TRANSFER TUBE HIGH CAPACITY
Supplier: 0000013443
HOBO® Temperature Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
Microliter™ and Gastight® Neuros™ Syringes, Hamilton
Supplier: Hamilton
PROTEIN DIGESTION KIT MINI ACCUMAP LOWPH
Supplier: Promega Corporation
VWR® Upright Freezer Racks for 96 Deep Well Microtiter Plates, Stainless Steel
Supplier: VWR International
ZORBAX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
NEEDLE INSERT HESI METAL STANDARD FLOW
Supplier: THERMO ELECTRON NORTH AMERICA MS
ENZYME GLYSERIAS 2000 UNITS
Supplier: GENOVIS, INC MS
Alpha 2-3,6,8,9 Neuraminidase A
Supplier: New England Biolabs (NEB)
KIT SYRINGE ADAPTER
Supplier: THERMO ELECTRON NORTH AMERICA MS
O-RING FOR APERTURE PK10
Supplier: 0000013719