About this item
Papain dissociation system is a set of reagents intended for use in the neural cell isolation method of Huettner and Baughman. Papain is used with fetal as well as postnatal brain regions to provide maximal dissociation and viability of neurons.
- Convenient and simple
- Provides freshly prepared enzyme solutions for each dissociation
- Useful to the occasional user as well as the more experienced and frequent user
Designed for the convenient isolation of single, morphologically intact cells for cell culture, flow cytometry or other applications.
For larger tissue samples prepare proportionately larger volumes of reagents at each step and combine them in the same ratio as described in the protocol. Individual component vials are also available for modified preparations in addition to a complete kit without EBSS for specialized buffer requirements.