ContactUsLinkComponent
Stopcock, Three Way, Radnoti
Catalog # MSPP-120721-00
Supplier:  AD INSTRUMENTS MS
Stopcock, Three Way, Radnoti
Catalog # MSPP-120721-00
Supplier:  AD INSTRUMENTS MS
Supplier Number:  120721-001
Specifications
  • Description:
    Three-way stop cock with Luer lock
  • Cat. No.:
    MSPP-120721-00
  • Supplier no.:
    120721-001
Specifications
About this item
This unique three-way micro valve is ideally designed for controlling the flow of liquid or gas. The use of this valve permits sterilizing and removes a source of metal contamination and corrosion that occurs using metal fittings.
  • Polycarbonate construction with polyethylene handle
  • Two female and one male outlet
  • Autoclavable and provide quick, leak-tight connections
This valve is mainly used in the making of manifolds to switch gases, to remove fluid samples from organ baths or to act as fittings for pressure transducers.
Ordering information: Male and female Luer connectors and adapters are also available.