ContactUsLinkComponent
Stackable Beakers, United Scientific Supplies
Stackable Beakers, United Scientific Supplies
Catalog # 76263-398
Supplier #: BST500-CASE
undefined
Stackable Beakers, United Scientific Supplies
Catalog # 76263-398
Supplier Number:  BST120-CASE
Supplier #: BST500-CASE
Log in for availability
Select an Option
108 mm
Select an Option
50 ml
Select an Option
50 - 500 ml
Select an Option
500 ml
Select an Option
95 mm
Select an Option
76263-398
About this item
Flexible design for pouring and stacking.
  • Stackable
  • Autoclavable
These autoclavable Polypropylene beakers are highly flexible, allowing a gentle squeeze to create a spout anywhere on the rim. Beakers feature a large writing area and are graduated in both ounces and milliliters. Stacking the ribs prevents the beakers from sticking together.
Frequently Bought Together