Order Entry
United States
Orders LinkContactUsLinkComponent
SuperFrost Plus® Microscope Slides

SuperFrost Plus® Microscope Slides

Glass slides, Superfrost, White, 1.1 mm, 75×25 mm, 144/Pack, 10/Case
Catalog # MSPP-1255015
Marketsource
Supplier #:  12-550-15
SuperFrost Plus® Microscope Slides
Catalog # MSPP-1255015
Marketsource
SuperFrost Plus® Microscope Slides

Glass slides, Superfrost, White, 1.1 mm, 75×25 mm, 144/Pack, 10/Case

Price
$1,555.71
Log in for availability
Case of 10
Minimum order quantity is 1
Product Details & Documents

This microscope slide is specially designed for the treatment process that electrostatically adheres tissue sections and cells to glass.

  • Chemical-resistant white marking surface
Specifications
  • Color:
    White
  • Description:
    Glass slides, Superfrost
  • L×W:
    75×25 mm
  • Thickness:
    1.1 mm
  • Packaging:
    144/Pack, 10/Case
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.