Order Entry
United States
ContactUsLinkComponent
Primocin™, Invivogen
Primocin™, Invivogen
Supplier:  Invivogen
Certificates
Primocin™, Invivogen
Supplier:  Invivogen
Certificates

Specifications

  • Endotoxin:
    <0.5 EU/ml
  • Storage Temperature:
    –20 °C
  • Concentration:
    50 µg/ml

Specifications

Related Information

Frequently Bought Together