C-Flex® Tubing, Cole Palmer
Supplier: Avantor
Certificates
About this item
C-Flex® is a clear, transparent tubing, suitable for the pharmaceutical, biotechnology and general-purpose lab uses requiring high performance.
- Economic alternative for platinum silicone
- Exceptional tensile and tear strength
- Low gas permeability
- Smooth surface
- Good biocompatibility
- Excellent chemical resistance
Temperature ranges from –45 to +275 °C. Sterilization can be done through autoclave, ethylene dioxide or gamma irradiation.
Certifications: Manufactured in accordance with Part 820 Quality System Requirements (GMP) and complies with USP Class VI standards.
Frequently Bought Together