ContactUsLinkComponent
HPLC Column Heater, Cole Palmer
HPLC Column Heater, Cole Palmer
Catalog # MSPP-UX4265070
Supplier:  Cole Parmer
undefined
HPLC Column Heater, Cole Palmer
Catalog # MSPP-UX4265070
Supplier:  Cole Parmer
Supplier Number:  42650-70
Specifications
  • Description:
    HPLC Column Heater
  • Cat. No.:
    MSPP-UX4265070
  • Electrical:
    90 - 260 VAC, 50/60 Hz
Specifications
About this item
Suitable for the columns that are 1/4" or 3/8" diameter by up to 30 cm long.
  • Compact
  • Easy-to use
  • Features up and down arrow for temperature set-up
  • With 30-days count down timer
  • Features platinum RTD temperature sensor and PID temperature controller
The silicon rubber heater underneath the chamber provides uniform chamber temperatures for optimal results. The chamber design provides quick access to the column and features a clear polycarbonate cover for easy viewing. Target and actual temperatures will be displayed—the stable temperature light will illuminate once the target temperature is reached and settled to within ±0.2 °C.