Order Entry
United States
ContactUsLinkComponent
Oakton® PCTSTestr™ 5 Waterproof Pocket Tester, Cole-Parmer
Oakton® PCTSTestr™ 5 Waterproof Pocket Tester, Cole-Parmer
Catalog # 76258-462
Supplier:  Antyila Scientific
undefined
Oakton® PCTSTestr™ 5 Waterproof Pocket Tester, Cole-Parmer
Catalog # 76258-462
Supplier:  Antyila Scientific
Supplier Number:  35634-60

Specifications

  • Calibration:
    1 − 3 points
  • Temperature Range:
    0...50 °C (32...122 °F)
  • Description:
    Waterproof Pocket Multiparameter Tester; pH, Conductivity, TDS, Salinity, and Temperature
  • Temperature Accuracy:
    ±0.5 °F/°C
  • Temperature Resolution:
    0.1 °F/°C
  • Battery Life:
    400 hours
  • Conductivity Accuracy:
    ±1% full scale
  • Conductivity Range:
    0.00 - 20.00 mS/cm
  • Conductivity Resolution:
    0.1 µS/cm, 1 µS/cm, 0.01 mS/cm
  • Display:
    Dual-line LCD
  • Salinity Accuracy:
    ±1% full scale
  • Salinity Range:
    0.0 − 10.0 ppt
  • Salinity Resolution:
    0.01 ppt
  • TDS Accuracy:
    ±1% full scale
  • TDS Range:
    0.1 − 10.0 ppt
  • TDS Resolution:
    0.1 ppm, 1 ppm, 0.01 ppt
  • Temperature Compensation:
    Automatic
  • pH Accuracy:
    ±0.01 pH, ±1 digit
  • pH Range:
    –2 – 16
  • pH Resolution:
    0.01 pH
  • Cat. No.:
    76258-462

Specifications

About this item

Save money by replacing sensor, not tester!

  • Conductivity, pH, salinity, and TDS all in one meter
  • 3-point calibration with automatic recognition of standards
  • Large easy-to-read LCD features two-color backlight
  • Rugged housing is waterproof, dustproof, and floats in water when sensor cap is on
  • Leakproof cap with fill line functions as a sample/calibration and hydrating cup
  • Two years warranty

NIST and USA buffer recognition.

Certifications: CE marked, IP 67 rated housing.
Delivery: Includes plastic storage case, lanyard, and four AAA batteries.