Order Entry
United States
ContactUsLinkComponent
 
Oakton® EcoTestr™ pH1 Waterproof Pocket Tester, Cole-Parmer
Catalog # 76258-452
Supplier:  Antyila Scientific
undefined
Oakton® EcoTestr™ pH1 Waterproof Pocket Tester, Cole-Parmer
Catalog # 76258-452
Supplier:  Antyila Scientific
Supplier Number:  35634-05

Specifications

  • Calibration:
    1 − 3 points
  • Temperature Range:
    0...50 °C (32...122 °F)
  • Description:
    Waterproof pH Tester
  • Temperature Accuracy:
    ±0.9 °F (±0.5 °C)
  • Temperature Resolution:
    0.1 °F/°C
  • Battery Life:
    400 hours
  • Display:
    LCD
  • W×D×H:
    40×31×178 mm (1.5×1.2×7")
  • pH Accuracy:
    ±0.1 pH
  • pH Range:
    0 − 14
  • pH Resolution:
    0.1 pH
  • Cat. No.:
    76258-452

Specifications

About this item

Automatically recognizes pH buffer standards.

  • Large LCD dual-line display shows pH and temperature readings simultaneously
  • Rugged housing is waterproof, dustproof, and floats in water
  • Leakproof cap with fill line functions as a sample/calibration and hydrating cup
  • Secure - attach a lanyard to prevent losing or dropping in the field
  • Auto shutoff extends battery life
  • Two years warranty

NIST and USA buffer recognition.

Certifications: CE marked, IP 67 rated housing.
Delivery: Includes plastic storage case, lanyard, and four AAA batteries.