Specifications
- Description:QuantiChrom™ β-N-Acetylglucosaminidase Assay Kit
- Size:100 assays
- Cat. No.:76237-112
Specifications
About this item
β-N-Acetylglucosaminidase activity determination in biological samples.
- Fast and sensitive, linear detection range (20 µl sample): 0.2 to 50 U/l for a 30 minute reaction at 37 °C
- High-throughput
- Homogeneous "mix-incubate-measure" type assay
- Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
- Method: OD405 nm
- Samples: Urine, serum, plasma, cell lysate, and more
β-N-Acetylglucosaminidase (NAG) is a lysosomal enzyme involved in a variety of biological processes such as the degradation of glycoproteins and glycolipids, cell proliferation, and signal transduction. NAG is found in many tissues in the body, but due to its high molecular weight, it can not be filtered through the glomerular membrane. For this reason, in the presence of tubular damage or a glomerular lesion, urinary NAG activity increases. Elevated NAG levels in urine are an early indication of renal damage, such as injury due to diabetes mellitus, inflammation, nephritic syndrome, urinary tract infection, and more. Various forms of cancer have been associated with increased levels of NAG in serum. Genetically inherited lipid storage disorders, such as Tay-Sachs and Sandhoff disease, arise from deficiencies of the enzyme. BioAssay Systems non-radioactive, colorimetric NAG assay is based on the cleavage of p-nitrophenol from a synthetic substrate. p-Nitrophenol becomes intensely colored after addition of the stop reagent. The increase in absorbance at 405 nm after addition of the stop reagent is directly proportional to the enzyme activity.