Order Entry
United States
ContactUsLinkComponent
QuantiChrom™ Peroxidase Assay Kit, BioAssay Systems
QuantiChrom™ Peroxidase Assay Kit, BioAssay Systems
Catalog # 76237-096
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiChrom™ Peroxidase Assay Kit, BioAssay Systems
Catalog # 76237-096
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  D2PD-100
CAS Number:  

Specifications

  • Description:
    QuantiChrom™ Peroxidase Assay Kit
  • Size:
    100 assays
  • Cat. No.:
    76237-096

Specifications

About this item

For quantitative determination of peroxidase activity.

  • Fast and sensitive, convenient and high-throughput
  • Use as little as 10 μl samples
  • Linear detection range: colorimetric assays 2 to 50 U/l, fluorimetric assays 0.1 to 5 U/l peroxidase
  • Homogeneous "mix-incubate-measure" type assay
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
  • Method: OD570 nm, or FL530/585 nm
  • Samples: Biological

Peroxidase (EC number 1.11.1.x) catalyze the following oxidation-reduction reactions:
ROOR + electron donor (2 e-) + 2H+ → ROH + R’OH. For many peroxidases the optimal substrate is hydrogen peroxide (H₂O₂), but others are more active with organic hydroperoxides such as lipid peroxides. In the cell, peroxidases destroy toxic hydroxide radicals that are formed as byproducts during aerobic respiration. The peroxidases represent a large family of enzymes that are found in animals (e.g. myeloperoxidase-like enzymes), plant, fungi and bacteria (cytochrome-c peroxidase like enzymes such a horseradish peroxidase). Simple, direct and automation-ready procedures for determining peroxidase activity find wide applications. Peroxidase assay uses H₂O₂ and an electron donor dye that forms a pink color during the peroxidase reaction. The optical density (570nm) or fluorescence intensity (λex/em = 530/590nm) is a direct measure of the enzyme activity.