Order Entry
United States
ContactUsLinkComponent
QuantiChrom™ Lactate Dehydrogenase Kit, BioAssay Systems
QuantiChrom™ Lactate Dehydrogenase Kit, BioAssay Systems
Catalog # 76237-094
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiChrom™ Lactate Dehydrogenase Kit, BioAssay Systems
Catalog # 76237-094
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  D2DH-100
CAS Number:  

Specifications

  • Description:
    QuantiChrom™ Lactate Dehydrogenase Kit
  • Size:
    100 assays
  • Cat. No.:
    76237-094

Specifications

About this item

For quantitative determination of lactate dehydrogenase LDH activity and screen or evaluation of LDH modulators.

  • High sensitivity and wide linear range, use 3 μL serum or plasma sample
  • The detection limit is 2 U/l, linear up to 200 U/l, homogeneous and simple procedure
  • Simple "mix-and-measure" procedure allows reliable quantitation of LDH activity within 30 minutes
  • Robust and amenable to HTS
  • All reagents are compatible with high-throughput liquid handling instruments
  • Method: OD565 nm
  • Samples: Serum, plasma and more

Lactate dehydrogenase (LDH) is an oxidoreductase which catalyzes the interconversion of lactate and pyruvate. When disease or injury affects tissues containing LDH, the cells release LDH into the bloodstream, where it is identified in higher than normal levels. Therefore, LDH is most often measured to evaluate the presence of tissue or cell damage. The non-radioactive colorimetric LDH assay is based on the reduction of the tetrazolium salt MTT in a NADH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The intensity of the purple color formed is directly proportional to the enzyme activity.