ContactUsLinkComponent
QuantiQuik™ L-Glutamate Quick Test Strips, BioAssay Systems
QuantiQuik™ L-Glutamate Quick Test Strips, BioAssay Systems
Catalog # 76236-152
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiQuik™ L-Glutamate Quick Test Strips, BioAssay Systems
Catalog # 76236-152
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  QQGLUT10
CAS Number:  
Specifications
  • Description:
    QuantiQuik™ L-Glutamate Quick Test Strips
  • Size:
    10 assays
  • Cat. No.:
    76236-152
Specifications
About this item
Direct determination of L-Glutamate (monosodium glutamate or MSG) concentrations in food and beverage samples as well as biological samples.
  • Fast and sensitive
  • Use of 20 or 100 µl sample
  • Semi-quantitative measurement between 0-300 mg/l (undiluted) L-Glutamate
  • Convenient. No extra equipment required
  • Sample treatment and assay can be performed in under 15 minutes
  • Method: Visual
  • Samples: Soups, sauces, milk, and more
Glutamate, is an important chemical in general metabolism. It is also widely used as a flavor enhancer in the food industry. Consumption of foods containing monosodium glutamate (MSG) have been known to cause headaches, flushing, sweating, nausea, and more. Glutamate is also a crucial mammalian neurotransmitter that is believed to be involved in a number of neurological and psychiatric disorders such as lateral sclerosis, autism, and Alzheimer’s disease.
QuantiQuik™ L-Glutamate Quick Test Strips are based on Glutamate dehydrogenase catalyzed oxidation of glutamate in which the formed NADH reduces a chromogenic reagent. The intensity of the product color is directly proportional to the glutamate concentration in the sample.