QuantiQuik™ Glucose Quick Test Strips, BioAssay Systems
Catalog # 76236-150
Supplier: BIOASSAY SYSTEMS
CAS Number:
Specifications
- Description:QuantiQuik™ Glucose Quick Test Strips
- Size:10 assays
- Cat. No.:76236-150
Specifications
About this item
Direct determination of glucose concentrations in food and beverage samples as well as biological (example: urine, blood, and more.) samples.
- Fast and sensitive
- Use of 20 or 100 µl sample
- Semi-quantitative measurement between 0-720 mg/l (undiluted) Glucose
- Convenient. No expensive lab equipment needed.
- Sample treatment and assay can be performed in under 15 minutes
- Method: Visual
- Samples: Wine, beer, juice, and more
Glucose (C₆H₁₂O₆) is found in the food industry as either a natural component of the food or as an additive. Not only is glucose added to food and beverages for its impact on taste, texture, and color, but also because it can serve as a preservative.
QuantiQuik™ Glucose Test Strips are based on Glucose dehydrogenase catalyzed oxidation of glucose in which the formed NADH reduces a chromogenic reagent. The intensity of the product color is directly proportional to the glucose concentration in the sample.