ContactUsLinkComponent
QuantiQuik™ D-Lactic Acid Quick Test Strips, BioAssay Systems
QuantiQuik™ D-Lactic Acid Quick Test Strips, BioAssay Systems
Catalog # 76236-148
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiQuik™ D-Lactic Acid Quick Test Strips, BioAssay Systems
Catalog # 76236-148
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  QQDLAC10
CAS Number:  
Specifications
  • Description:
    QuantiQuik™ D-Lactic Acid Quick Test Strips
  • Size:
    10 assays
  • Cat. No.:
    76236-148
Specifications
About this item
Direct determination of D-lactic acid concentrations in food and beverage samples as well as biological samples.
  • Fast and sensitive
  • Use of 20 or 100 µl sample
  • Semi-quantitative measurement between 0-180 mg/l (undiluted) D-lactic acid
  • Convenient, no expensive lab equipment needed
  • Sample treatment and assay can be performed in under 15 minutes
  • Method: Visual
  • Samples: Wine, beer, milk, and more
Lactic acid, or lactate, is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions. D-lactate is produced in only minor quantities in animals and measuring for D-lactate in animal samples is a means to determine the presence of bacterial infection. Furthermore, since D-lactate is a specific indicator of bacteria fermentation, its measurement can be used to determine the freshness of milk, meat and fruit juices. Elevated levels of D-lactate in wine is an indication of lactic acid bacteria contamination.
QuantiQuik™ D-Lactate Test Strips are based on D-Lactate dehydrogenase catalyzed oxidation of D-lactate in which the formed NADH reduces a chromogenic reagent. The intensity of product color, is directly proportional to D-lactate concentration in the sample.