ContactUsLinkComponent
EnzyFluo™ Tryptophan Assay Kit, BioAssay Systems
EnzyFluo™ Tryptophan Assay Kit, BioAssay Systems
Catalog # 76236-158
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyFluo™ Tryptophan Assay Kit, BioAssay Systems
Catalog # 76236-158
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  ETRP-100
CAS Number:  
Specifications
  • Description:
    EnzyFluo™ Tryptophan Assay Kit
  • Size:
    100 assays
  • Cat. No.:
    76236-158
Specifications
About this item
Direct determination of tryptophan in serum.
  • Fast and sensitive, Ccnvenient
  • Use of 10 µl sample
  • Linear detection range from 10µM to 400 µM tryptophan in 96-well plate assay
  • Room temperature assay, no 37 °C heater is needed
  • High-throughput,"Add-mix-read" type assay. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: FL530/585 nm
  • Samples: Serum
The procedure involves adding a single working reagent, and reading the fluorescence after 30 minutes. Tryptophan is one of the eight essential amino acids that the body cannot synthesize and must be obtained through diet. Tryptophan is the biochemical precursor to the neurotransmitter serotonin, which has important roles in biological processes such as regulation of appetite, sleep, and mood. Imbalances of serotonin have been linked to numerous mental health disorders. Tryptophan is also a precursor to the neurotransmitter melatonin, which is heavily involved in regulating the body’s sleep cycle. EnzyFluo™ Tryptophan Assay uses a coupled enzymatic reaction to determine the tryptophan concentration of a sample with the addition of a single working reagent. The fluorescence intensity at λex/em = 530/585 nm is directly proportional to tryptophan concentration in the sample.