ContactUsLinkComponent
QuantiFluo™ Cholesterol Uptake Assay Kit, BioAssay Systems
QuantiFluo™ Cholesterol Uptake Assay Kit, BioAssay Systems
Catalog # 76237-102
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiFluo™ Cholesterol Uptake Assay Kit, BioAssay Systems
Catalog # 76237-102
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DCUT-100
CAS Number:  
Specifications
  • Description:
    QuantiFluo™ Cholesterol Uptake Assay Kit
  • Size:
    100 assays
  • Cat. No.:
    76237-102
Specifications
About this item
Cholesterol uptake by adherent cells, screening of cholesterol uptake inhibitors, and evaluation of effect of drugs on cholesterol uptake.
  • Sensitive and accurate
  • Detection limit: 0.3 U/L for 2 hr arginase reaction in 96-well assay format
  • Safe. Non-radioactive assay
  • High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: FL485/535 nm
  • Samples: Adherent cells
Cholesterol is a sterol and lipid present in cell membranes, and is transported in the bloodstream of all animals. It is used to form cell membranes and hormones, and plays important roles in cell signaling processes. Cellular regulation of cholesterol levels is a complex system in which irregularities have been tied to obesity and heart disease. Increased cholesterol uptake has also been linked to highly proliferative cancer cells. Through monitoring cellular cholesterol uptake, one can explore these growing health problems and screen for possible drug treatments.
Cholesterol uptake assay kit is based on cellular uptake of a fluorescently tagged cholesterol probe. The fluorescence intensity measured at λex/em = 485/535 nm is proportional to the amount of cholesterol uptaken by the cells.