Synthetic Broth, AOAC (Wright and Mundy Broth), HiMedia Laboratories
: HiMedia
Synthetic Broth, AOAC is a chemically defined medium recommended by AOAC for growing inoculum, making subcultures and preparing various dilutions while testing disinfectants.
Frequently Bought Together