Order Entry
United States
ContactUsLinkComponent
 
Sabouraud Dextrose Broth, General Laboratory Products
Catalog # 101205-638
Minority or Woman-Owned Business Enterprise
CAS Number:  
Sabouraud Dextrose Broth, General Laboratory Products
Catalog # 101205-638
Supplier Number:  8WM-3300-100
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, wide mouth, 8 oz.
  • Description:
    Sabouraud Dextrose Broth, 12 per case
  • Size:
    100 ml
  • Cat. No.:
    101205-638
  • Supplier no.:
    8WM-3300-100

Specifications

About this item

Base for the cultivation of fungi and aciduric microorganisms.