ContactUsLinkComponent
Protein A ELISA Kit, Cygnus Technologies
Protein A ELISA Kit, Cygnus Technologies
  102650-526
 :  Cygnus
 :  
Protein A ELISA Kit, Cygnus Technologies
  102650-526
 :  Cygnus
 :  F400
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Protein A ELISA Kit
  • Size:
    1 Kit
  • Cat. No.:
    102650-526
  • Supplier no.:
    F400
Through the new method Mix-N-Go in protein A ELISA kit eliminate the sample boiling and centrifugation steps instead, a special sample treatment reagent is used that allows for accurate detection of Protein A in the presence of your sample antibody.
  • Improved detection of newer, unnatural constructs of protein A.
 : Kit includes anti-protein A:HRP conjugate, protein A standards set, A-G, sample denaturing buffer, sample treatment plate, stop solution and TBS wash concentrate, 20X.