Rocking Shaker, G-Biosciences
Catalog # 76264-012
Supplier: G-Biosciences
Irregular Voltage
Specifications
- Description:Rocking Shaker
- Max. Load:5 kg
- Weight capacity:5 kg
- Dimensions:284×264×100 mm
- Material:Stainless steel
- Operating Temperature:4...45 °C
- Timer:1 min. to 99 h, 59 min
- Cat. No.:76264-012
- Electrical:100/240 V, 1.5 A
Specifications
About this item
The rocking shaker is used in immobilization when running gels, shaking for dyeing or fading Coomassie blue, immobilization, dyeing and developing of silver-staining.
- Speed and time display
- Stable with no wear, suitable for prolonged use
- Compactly designed and takes up very little space
- Brushless DC motor, maintenance-free
It is also used in developing and fixation of X-ray film in autoradiography experiments and processing of cellulose membranes after gel shift such as molecular hybridization and staining with antigen-antibody. Cell culture and cell membrane transfer are uses for it also.
Platform is made of stainless steel and natural rubber, which is alkali resistant, non-deforming, easy to clean and prevents spills.