ContactUsLinkComponent
Acid Fast Stain Kit, Kinyoun, Cold Method for AFB, Hardy Diagnostics
Acid Fast Stain Kit, Kinyoun, Cold Method for AFB, Hardy Diagnostics
Catalog # 76213-940
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Acid Fast Stain Kit, Kinyoun, Cold Method for AFB, Hardy Diagnostics
Catalog # 76213-940
Supplier:  Hardy Diagnostics
Supplier Number:  AF900
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    Acid Fast Stain Kit
  • Size:
    3×250 ml
  • Cat. No.:
    76213-940
Specifications
About this item
The kit is used as a differential method to detect the difference between those organisms which decolorize with treatment of acid alcohol, and those that do not, such as the mycobacteria.
This is the “cold stain” method that does not require heating of the slide.
Delivery: Kit includes one 8 oz. bottle of each, carbol fuchsin (kinyoun), acid alcohol (decolorizer) and methylene blue (counterstain).