To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:HardyCHROM Salmonella/XLT-4 agar biplate
- Dimensions:15×100 mm (Plate)
- Minimum Order Quantity:20 packs
- Cat. No.:76220-924
- Fill volume:10 ml
- Storage temperature:2...8 °C
- Application:Food safety
Specifications
About this item
XLT-4 agar is a highly selective plating medium for the detection and isolation of non-typhi Salmonella species.
HardyCHROM™ Salmonella agar is a chromogenic medium recommended for the selective isolation and differentiation of Salmonella species from other members of the family Enterobacteriaceae based on colony color.
pH - HardyCHROM™ Salmonella: 7.2 ±0.2 at 25 °C; XLT-4 agar: pH 7.4 ±0.2 at 25 °C