Magnetic Stirrer, Digital, G-Biosciences
Catalog # 76263-980
Supplier: G-Biosciences
Irregular Voltage
Specifications
- Temperature range:5…40 °C
- Description:Magnetic stirrer, digital
- Environmentally Preferable:
- Speed Range:300 - 1200 rpm
- W×D×H:270×156×70 mm
- Max. Stirring Capacity H₂O:20 L
- Plate Dimensions:154×163 mm
- Plate Material:Silicone
- Power input/output:30 W (Input)
- Cat. No.:76263-980
- Electrical:100/230 V, 50/60 Hz, 1.5 A
- Weight:2.6 kg
Specifications
About this item
Digital magnetic stirrer designed with stir technology using a stirrer bar. It is used in medical science, bioengineering, chemistry, medicine and foodstuff. It is a laboratory instrument for stirring and mixing of various reagents, solutions and chemical substances.
- For stirring standard/non-standard reaction flasks between 50 and 1000 ml
- DC brushless motor, stable operation, low noise, with no sparking
- Metal plate shell is durable, has stable temperature and is corrosion resistant
- Suitable for use at 5 to 40 °C
Magnetic stirring technology and has steady operation at low speed, powerful operation at high speed. Digital LED display to control the panel designed on a 30° slopes for convenient operation.
Caution: Do not wall up or cover the vent.