ContactUsLinkComponent
DNAse I Activity Assay Kit (Fluorometric), BioVision, Inc.
DNAse I Activity Assay Kit (Fluorometric), BioVision, Inc.
Catalog # 76264-670
Supplier:  Avantor
CAS Number:  
undefined
DNAse I Activity Assay Kit (Fluorometric), BioVision, Inc.
Catalog # 76264-670
Supplier:  Avantor
CAS Number:  
Specifications
  • Description:
    DNAse I Activity Assay Kit (Fluorometric)
  • Size:
    100 assays
  • Cat. No.:
    76264-670
Specifications
About this item
High-throughput compatible, quick and easy screening for DNAse activity in biological samples
  • Quantitative analysis of DNAse I mutants and inhibitors
Deoxyribonuclease I (DNAse I) is an endonuclease that cleaves DNA phosphodiester bonds yielding 5′-phosphorylated and 3′-hydroxylated oligonucleotides. DNAse I targets single-stranded DNA, double-stranded DNA, and chromatin in a non-specific manner. As an important player in cellular waste management, DNAse I is normally secreted extracellularly to clear the system from circulating cell-free DNA, foreign DNA from food digestion or potential pathogens, and endogenous chromosomal DNA from apoptotic and necrotic cells. Abnormal DNAse I activity occurs in association with a variety of cancers and auto-immune illnesses that exhibit elevated levels of cell-free DNA. Furthermore, DNAse I has been therapeutically used in cystic fibrosis patients to degrade DNA and reduce sputum viscosity. BioVision’s DNAse I Activity Fluorometric Assay Kit allows for quantitative evaluation of DNAse I activity of purified enzymes and their inhibitors as well as comparative examination of DNAse I activity in biological samples. Enzyme activity is detected upon cleavage of a DNA Probe, which yields a fluorescent DNA product measured at Ex/Em = 651/681 nm. The limit of quantification (L.O.Q) is 178 fmoles of DNA probe cleaved per minute per ml