To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Complement C3
- Antigen Symbol:C3
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:2 ml
- Format:Lyophilized IgG
- Form:Lyophilized powder
- Antibody Fragment:F(ab')2
- Storage Buffer:pH: 7.3
- Storage Temperature:Store lyophilized product at 2-8°C. For long-term storage, aliquot and freeze reconstituted product at –20 °C or lower.
- Concentration:Total Protein: 28.8 mg/mL; Titer: 1:64,000
- Shipping Temperature:4C
- Purification:Salt fractionation
- Cat. No.:75983-040
Specifications
About this item
Product is the lyophilized powder of horseradish peroxidase (HRP)-conjugated goat IgG fraction to human complement C3 and
buffer salts.
Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: Horseradish Peroxidase (HRP)
Epitope:
Host: Goat
Isotype: Polyclonal IgG
Reactivity: Human