Order Entry
United States
ContactUsLinkComponent
Creatinine Deiminase (from Microbial), MP Biomedicals
Creatinine Deiminase (from Microbial), MP Biomedicals
Catalog # 76204-114
Supplier:  MP Biomedicals
CAS Number:  
Creatinine Deiminase (from Microbial), MP Biomedicals
Catalog # 76204-114
Supplier:  MP Biomedicals
Supplier Number:  0215349025
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle, amber
  • CAS No.:
    37289-15-9
  • Source:
    Microbial
  • Species:
  • Size:
    25 units
  • Environmentally Preferable:
  • Storage Conditions:
    −20 °C
  • Form:
    white powder
  • MDL No.:
    MFCD00130878
  • Enzyme Name:
    Creatinine Deiminase
  • Enzyme Activity:
    ≥10 U/mg solid
  • Cat. No.:
    76204-114

Specifications

About this item

  • Presentation : White Powder
  • Isoelectric point (pI) : 4.4 (Lit.)
  • pH : Optimum pH: 8.5 – 9.5 (Lit.)
  • pH Stability : pH 7.0 – 11.0 (30 °C, 20hr)
  • NOTES : Michaelis constant : 3.5 x 10¯3M (Creatinine), Thermal stability : Below 65 °C (pH 7.5, 1hr), Optimum temperature : 65 – 75 °C, (Lit.)