Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:5 - FITC Protein Labeling Kit AnaTag™ 'Ultra Convenient'
- Size:1 kit
- Storage Conditions:4°C
- Shipping Conditions:20
- Cat. No.:103010-374
- Abs/Em:494/519 nm
- Supplier no.:AS-72059
Fluorescein isothiocyanate (FITC) remains one of the most popular fluorescent labeling reagents, probably due to the low cost of the material. 5-FITC and 6-FITC have very similar absorption and fluorescence spectra. However, the isomers may differ in their binding and reactivity to proteins, and the conjugates may elute under different chromatographic conditions or migrate differently in electrophoretic gels.
5-FITC is more widely used than 6-FITC isomer for protein labeling. FITC-protein conjugates are suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. The kit has all the essential components for performing the conjugation reaction and for purifying the FITC-protein conjugates. The kit is good for three conjugation reactions with each reaction labeling up to 5 mg IgG. Kit Size: 3 labeling reactions (3 x 5 mg protein)