Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Fluor 750 Protein Labeling Kit AnaTag™ HiLyte™ 'Ultra Convenient'
- Size:1 kit
- Storage Conditions:4°C
- Shipping Conditions:20
- Cat. No.:103010-346
- Abs/Em:754/778 nm
- Supplier no.:AS-72043
Specifications
About this item
HiLyte™ Fluor 750 acid, SE is the longest-wavelength amine-reactive HiLyte™ Fluor dye currently available. Its fluorescence emission maximum at 778 nm is well separated from commonly used far-red fluorophores such as HiLyte™ Fluor 647, HiLyte™ Fluor 680 or allophycocyanin (APC), facilitating multicolor analysis.
With a peak excitation at ~754 nm, conjugates of HiLyte™ Fluor 750 dyes are well excited by a xenon-arc lamp or dye-pumped lasers operating in the 720–750 nm range. The HiLyte™ Fluor 750-protein conjugates are suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. The kit has all essential components for performing the conjugation reaction and for purifying the HiLyte™ Fluor 750-protein conjugates. The kit is able to perform three conjugation reactions with each reaction labeling up to 5 mg IgG. Kit Size: 3 labeling reactions (3 x 5 mg protein)