Y-27632 dihydrochloride, InSolution™, Calbiochem®
Catalog # 80512-706
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Size:500 µg
- Cat. No.:80512-706
- Supplier no.:688001-500UG
Specifications
Specification Test Results
- Primary Target:p160ROCK
- Primary Target IC50:700 nM against phenylephrine-induced contractions in aortic strips
- Primary Target Ki:140 nM for p160ROCK (ROCK-I)
- Purity:≥95% by HPLC
Specification Test Results
About this item
:
:320.26 g/mol
:Packaged under inert gas.
:GHS07
Storage Temperature:Freezer
CAS Number:129830-38-2 | 146986-50-7 (free base)
:146986-50-7
Formula:C₁₄H₂₁N₃O·2HCl
MDL Number:MFCD03490488