Fisherbrand Petri Dish, PANTek Technologies
Catalog # MSPP-FB0875713
Supplier: PANTEK TECHNOLOGIES MS
Specifications
- Description:Fisherbrand petri dish
- Size:100×15 mm
- Environmentally Preferable:
- Cat. No.:MSPP-FB0875713
- Supplier no.:FB0875713
Specifications
About this item
Made of polystyrene.
- Slippable
- Beveled ridge
Redesigned packaging (outer carton, strong polymer vacuum sealed sleeves) prevent scuffing and breakage. Inner sleeves feature easy-open tear notch and lot number.