Hemocytometer, Disposable, Diagnocine
MSPP-52110
- Description:Disposable hemocytometer (4-chambers), large
- Cat. No.:MSPP-52110
Disposable hemocytometer with 4 chambers, made of plastic.
- Designed for rapid loading
- No washing, no labor, saves time
- Same grid as standard hemocytometer
- High calculation accuracy