Order Entry
United States
ContactUsLinkComponent
Certified Reference Material, Nitric Acid Rinse Solution, Inorganic Ventures
Certified Reference Material, Nitric Acid Rinse Solution, Inorganic Ventures
Catalog # 103255-688
Supplier:  Inorganic Ventures
CAS Number:  
Certified Reference Material, Nitric Acid Rinse Solution, Inorganic Ventures
Catalog # 103255-688
Supplier:  Inorganic Ventures
Supplier Number:  CLPMSRINSE-125
CAS Number:  

Specifications

  • Concentration:
    2% (v/v)
  • Matrix:
    HNO₃
  • Description:
    Nitric Acid Rinse Solution
  • Size:
    125 ml
  • Cat. No.:
    103255-688
  • Supplier no.:
    CLPMSRINSE-125

Specifications

About this item

Nitric acid rinse is a reference material set in a nitric acid matrix for stability.

A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Certifications: Traceable to NIST and ISO 9001 certified.