Tissue-Tek® Cryomolds® Molds, Ted Pella Inc
Supplier: TED PELLA INC MS
Certificates
About this item
For use with Tissue-Tek® II M/C.
Tissue-Tek Cryomold® creates a uniformly shaped, flat-surface specimen blocks with O.C.T. Compound for frozen sectioning.
Disposable PVC plastic molds fit Cryobar® freezing wells.
Frequently Bought Together