PCR Vials, AB SCIEX
Catalog # MSPP-144709
Supplier: 0000013719
Specifications
- Description:PCR Vials
- Cat. No.:MSPP-144709
- Supplier no.:144709
Specifications
About this item
Microvials can be used as vial inserts for small volume sample introduction into the CESI 8000 Plus, PA 800series, and P/ACE™ MDQ systems.
Microvials fit into Universal vials.